missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Adjustable Angle Platform Rocker
Large capacity, adjustable angle and speed platform rocker, providing soft or intensive side to side rocking for optimal mixing of samples. Suitable for use in cold rooms or incubators with ambient operating temperature range +4ºC to 40ºC.
Brand: Grant Instruments™ PMR-100
1380.45 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
This item is not returnable.
View return policy
Description
- Tilt angle range, adjustable up to 10º
- Loads up to 5kg
- Variable speed: 1 to 99 oscillations per minute
- Continuous or timed operation, with automatic switch-off
- Smooth, non-slip mat supplied as standard
- Compact, sturdy construction with a low profile and small footprint

Specifications
0 to 10° | |
1 to 99 rpm | |
4°C to 40°C | |
480 mm | |
Adjustible Tilt |
5 kg | |
120/230 V | |
250 mm | |
400 mm | |
1 min. to 99:59 hr. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Grant Instruments™ Adjustable Angle Platform Rocker > Platform Style: Adjustable Tilt
Spot an opportunity for improvement?Share a Content Correction