Learn More
Nabertherm™ Compact Tube Furnace with B510 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
Brand:  Nabertherm™ R-472L3AN
 View more versions of this product
Product Code. 17388089
  • 10230.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Single-zoned design as standard with controller B 510
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Specifications
B510
1300°C
100 mm
750 mm
Compact Tube Furnace