missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Compact Tube Furnace R Series with P580 Controller

Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.

Brand:  Nabertherm™ R-572W3AN3

Product Code. 17388099

  • 13585.00 € / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Maximum Temperature: 1300°C
  • Single-zoned design as standard
  • Dual shell housing made of sheets of textured stainless steel
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Working tube of C 530 ceramic including two fiber plugs as standard equipment
  • Type S thermocouple
  • Solid state relays provide for low noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications

Specifications

P580
460 x 1170 x 628 mm
170 mm
1300°C
1000 mm
89 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers