Learn More
UVP Compact UV Lamp
Brand:  UVP 95-0019-14
 View more versions of this product
Product Code. 11738241
  • 168.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Compact UV Curing Lamp
4 W
198 x 71 mm
Plastic
71 mm
12 V
365 nm
198 mm
UVL-23R