missing translation for 'onlineSavingsMsg'
Learn More

Integra Biosciences™ Complete Hand Operator Kit

Convert any vacuum source into a user-friendly aspiration system. Integra Biosciences™ Vacuboy™ Complete Hand Operator Kit is a versatile aspiration tool for aspirating liquids from virtually every laboratory vessel.

Brand:  Integra Biosciences™ 155500

Product Code. 10125941

  • 595.00 € / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

VACUBOY is a versatile aspiration tool that converts any vacuum source into a user-friendly aspiration system for aspirating liquids from virtually any laboratory container at the touch of a button and in a controlled way.
  • Hand piece is connected to any available vacuum source using silicone tubing and fitted with one of the different aspiration adapters
  • The perfect match is obtained when VACUBOY is connected to a VACUSAFE aspiration system
  • The large choice of adapters allows VACUBOY to handle liquids from all tubes, dishes, bottles, T-flasks or multi-well plates, suiting their form and size
  • Adapters for aspiration with Pasteur pipettes, serological pipettes, disposable pipette tips or stainless steel needles
  • Multi-channel adapters for fast emptying of multi-well plates
  • Vacuum regulation by a touch-sensitive button for controlled and smooth aspiration of liquids
  • Ergonomic, perfectly suited for long working, productive or repetitive aspiration tasks
  • Anti-drip system prevents liquid waste leakage from the tip or pipette, reducing contamination risks when working with biohazardous liquids
  • Autoclavable and sterilizable, VACUBOY allows a maximal safety in the laboratory when working with hazardous biological liquid waste
  • Compact, user friendly, intuitive operation
  • Level detection sensor
  • Control knob for infinitely vacuum setting (range: -300mbar to 600mbar)
  • Silent operation
  • VACUBOY hand operator with 40mm stainless steel adapter included.
  • By turning the vacuum control knob to the desired level it will automatically control and adjust during operation. Stand alone or in combination with its many VACUBOY hand operator accessories, it is the perfect match for a wide range of vacuum based applications
  • All parts exposed to critical liquids are fully autoclavable and can be sterilized for 20 minutes at 121°C, guaranteeing maximum safety
  • Liquids are collected in the autoclavable waste bottle
  • Hydrophobic filter reduces the risk of accidently aspirating liquid into the pump and pump housing
  • Available with either a 4L polypropylene bottle or a 3L glass bottle.
  • VACUSAFE comfort instantly provides a clean solution for safe and convenient aspiration, disposal or filtration of liquids
Specifications

Specifications

Autoclavable
Stand and 6 Adapters (Rubber adapter for Pasteur pipettes, 1-Channel Stainless Steel adapter 40mm, 8-channel Stainless Steel adapter 40mm, 1-channel adapter for tips, 1-channel adapter for tips/discharge, 8-channel adapter for tips/shedding)
1 Each
Hand Operator Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.