missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Compound E Product Code.: 15426749

Thermo Scientific Chemicals Compound E

Product Code. 15426749
0.5 mg
Click to view available options
Quantity:
0.5 mg
This item is not returnable. View return policy

Product Code. 15426749

Brand: Thermo Scientific Chemicals J65131.!D

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 209986-17-4 | C27H24F2N4O3

A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of --secretase and Notch processing

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.5 mg
Description Off-white
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, potent, selective, peptidomimetic, non-transition-state analog inhibitor of →-secretase and Notch processing
CAS 209986-17-4
Molecular Formula C27H24F2N4O3
Formula Weight 490.50
Synonym γ-Secretase Inhibitor XXI;
Sensitivity Light Sensitive
Form Solid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY11K002 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ65131.!D
Lot NumberU02K009 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberJ65131.!D
Lot NumberR06K014 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog NumberJ65131.!D
Lot NumberU25J020 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberJ65131.!D
Lot NumberM23J022 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberJ65131.!D

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Compound E >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.