missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Con A Sepharose™ 4B Medium

An affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids

Brand:  Cytiva 17-0440-01


 View more versions of this product

Product Code. 11524905

This item is not returnable. View return policy

Description

Description

Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.

  • Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
  • Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Group-specific adsorbent for molecules containing sugars
  • Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
  • Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
  • Average particle size: 90μm
  • Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

30mg/mL
4% Agarose
4 to 9
2°C to 8°C
Concanavalin A
13mg Con A/mL Drained Medium
90 μm
100 mL
For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Con A Sepharose 4B, 100 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers