Learn More
Contec™ CONSTIX™ Surface Validation Swabs
BIOPHARMAGREENER_CHOICE
CONSTIX™ Surface Validation Swabs designed for the evaluation of surface cleaning protocols in biopharmaceutical industries.
14.70 € - 1728.00 €
Specifications
For Use With (Application) Evaluation of surface cleaning protocols in bio-pharmaceutical industries
Product Type Swab
Sterility Non-sterile
Adhesive Type Ultrasonically Sealed
Handle Material Polypropylene
View More Specs
Products 2
Product Code Brand Length (Metric) Tip Price Quantity & Availability  
Product Code Brand Length (Metric) Tip Price Quantity & Availability  
16685554
View Documents
Contec™
SV-2
2 cm N/A N/A N/A
16249721
BIOPHARMAGREENER_CHOICE
View Documents
Contec™
SV-3
2.31 cm
14.70 €
Pack of 5
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
Description
Description
  • Package contains five polyester swabs specially processed to minimize background TOC contribution from the swab
  • Low background TOC content for minimal interference with test results
  • Large surface of the swab head and the long flexible handle provide exceptional surface coverage when swabbing
  • Strong, durable polyester swabs for optimum retrieval
  • Sterilizable by most common methods
  • Autoclavable
  • Swabs sealed in nylon pouch
Specifications
Specifications
Evaluation of surface cleaning protocols in bio-pharmaceutical industries
Non-sterile
Polypropylene
Swab
Ultrasonically Sealed
Knit Polyester
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Contec™ CONSTIX™ Surface Validation Swabs