missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.

Brand:  Invitrogen™ C21421

241.50 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 11514247

  • 345.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Canavlia ensiformis
α-man, α-glc
Native
Store in freezer (-5 to -30°C) and protect from light.
Alexa Fluor Dyes
Natural
Lyophilized
Imaging
Alexa Fluor™
Lectin
5 mg
Alexa Fluor 647
C. ensiformis
None
650/668 nm
Room Temperature
Lectins
Concanavalin A (Con A)
Deep Red
Imaging Quality
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2860761 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog NumberC21421
Lot Number2720388 Certificate TypeCertificate of Analysis Date12/09/2023 Catalog NumberC21421
Lot Number2306777 Certificate TypeCertificate of Analysis Date19/03/2021 Catalog NumberC21421
Lot Number2273729 Certificate TypeCertificate of Analysis Date24/11/2020 Catalog NumberC21421
Lot Number2064010 Certificate TypeCertificate of Analysis Date04/02/2019 Catalog NumberC21421
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Concanavalin A (Con A) Conjugates > Alexa Fluor™ 647

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.