missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Concanavalin A (Con A) Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates.

Brand:  Invitrogen™ C11252

262.96 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 11594207

  • 346.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.

Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Flour and Alexa Flour Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immunocytochemistry, Immunohistochemistry, Immunofluorescence
5 mg
RUO
α-man, α-glc
Native
Store in freezer (-5 to -30°C) and protect from light.
Alexa Fluor Dyes
Natural
Lyophilized
Imaging
Alexa Fluor™
Lectin
104,000 Dalton
Canavlia ensiformis
Alexa Fluor 488
C. ensiformis
None
495/519 nm
Room Temperature
Lectins
Concanavalin A (Con A)
Green
Imaging Quality
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Manuals

Certificats

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2978757 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog NumberC11252
Lot Number2892448 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog NumberC11252
Lot Number2720398 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog NumberC11252
Lot Number2555720 Certificate TypeCertificate of Analysis Date24/03/2023 Catalog NumberC11252
Lot Number2540863 Certificate TypeCertificate of Analysis Date21/09/2022 Catalog NumberC11252
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Invitrogen™ Concanavalin A (Con A) Conjugates > Alexa Fluor™ 488

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis