missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Concentrator Plus
Eppendorf Concentrator plus was designed to deliver an ergonomic, intuitive and durable solution that fits comfortably right on your lab bench.
Brand: Eppendorf™ 5305000363
This item is not returnable.
View return policy
Description
Specifications
50/60 Hz | |
20 hPa (20 Mbar) | |
Concentrators | |
Concentrator Plus Complete with GB Plug | |
1400 rpm (max. RCF 250 x g) | |
58 x 33 x 29 cm (12.6 x 20.9 x 11.7 in.) |
With Rotor F-45-48-11 | |
Integrated diaphragm vacuum pump | |
230 V | |
230 V, 50/60 Hz (GB) | |
350 W | |
Rotor F-45-48-11 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction