Learn More
Motic™ Condenser for AE2000
Suitable condenser for AE2000 inverted microscopes. Motic™ Condenser for AE2000 has a short working distance.
Brand:  Motic™ 1101000202011
Product Code. 15455322
  • 682.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Concentrates and focuses light passing through the specimen.
  • Short working-distance condenser (28mm).
Specifications
Specifications
NA: 0.50
Condenser
AE2000 Microscopes
28 mm