Learn More
Lenz Laborglasinstrumente™ Condenser
Condenser for condensing vapors.
Brand:  Lenz Laborglasinstrumente™ 05210302
 View more versions of this product
Product Code. 12643830
  • 54.00 € / Each
Estimated Shipment: 16-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemical resistance
  • High temperature resistance
Specifications
Specifications
250 mm
NS 19/26
Condenser
1 Ea.
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Condenser > Neck Size: 19/26; Length: 250 mm