Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
117.00 € - 125.00 €
Specifications
Material Duran Glass
View More Specs
Products 2
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
10306171
View Documents
Lenz Laborglasintrumente
05226004
NS 29/32
117.00 €
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
10244101
View Documents
Lenz Laborglasintrumente
05226005
NS 34/35
125.00 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
Description
Description
  • Allihn condensers for condensing vapours
  • With NS cone
  • Water connection: olives
  • Made of DURAN™ tubing
Specifications
Specifications
Duran Glass