Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05215304
This item is not returnable.
View return policy
Description
- Davies condensers for condensing vapours
- With NS socket and NS cone
- With double sized cooling area
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Glass olives | |
250 mm | |
NS 29/32 | |
Borosilicate Glass (3.3) |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Davies > NS 29/32
Spot an opportunity for improvement?Share a Content Correction