Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies, with GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05215414
 View more versions of this product
Product Code. 11566794
  • 199.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Davies condensers for condensing vapours
  • With NS socket and NS cone
  • With double sized cooling area
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Specifications
Two threaded plastic olives
400 mm
NS 29/32
Duran Glass