Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05210714
This item is not returnable.
View return policy
Description
- Liebig condensers acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
GL 14 threads with two threaded plastic olives | |
750 mm | |
NS 29/32 |
750 mm | |
1 Ea. |
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Liebig with GL Threads > 750 mm
Spot an opportunity for improvement?Share a Content Correction