Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
75.00 € - 76.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | ||||||
10224631
|
Lenz Laborglasintrumente
05210401 |
NS 14/23 |
75.00 €
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10498592
|
Lenz Laborglasintrumente
05210403 |
NS 24/29 |
76.00 €
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- Condensers acc. to Liebig
- With olives Liebig condenser
- Acc. to DIN 12576
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) | |
400 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers Acc. to Liebig with Olives