Learn More
Lenz Laborglasintrumente Condensers acc. to Liebig, with Olives with Cone and Socket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05210404
Product Code. 10664392
  • 76.00 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liebig condenser acc. to DIN 12576
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Specifications
400 mm
Socket and Cone NS 29/32
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers acc. to Liebig, with Olives with Cone and Socket > 400 mm