missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Condensers with Cone and Socket
59.00 € - 63.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10159310
|
Lenz Laborglasinstrumente™
05210301 |
250 mm |
59.00 €
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10376071
|
Lenz Laborglasinstrumente™
05210202 |
160 mm |
63.00 €
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Specifications
Condenser | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lenz Laborglasinstrumente™ Condensers with Cone and Socket