missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Calibration Solution, 2764μS, Cole-Parmer™

500mL, Clear, None, 4.75 to 7.75, Liquid, Water Soluble, 0 to 10% KCl, NaCl, 90 to 100% Water

Brand:  Cole Parmer 00653-20

Product Code. 11779879

  • 18.65 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

500mL, Clear, None, 4.75 to 7.75, Liquid, Water Soluble, 0 to 10% KCl, NaCl, 90 to 100% Water

Specifications

Specifications

500mL
Calibration Solution
Conductivity Standard
0 to 10% KCl, NaCl, 90 to 100% Water
Soluble in water
500 mL
Clear
None
4.75 to 7.75
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.