Learn More
Mettler Toledo™ Conductivity Meter SevenDirect SD30

The SevenDirect SD30 is an accurate and flexible benchtop meter for the determination of conductivity, total dissolved solids and salinity. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
Brand: Mettler Toledo™ 30671563
1917.69 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.

Specifications
0.005 | |
0.001 to 1 | |
-22°F to +266°F | |
±0.1°C between 0°C and 100°C | |
2000 Standard Measurements | |
IP54 | |
850 g | |
InLab 741-ISM Conductivity/Temperature Sensor | |
SevenDirect SD30 Pure H2O Kit |
0.001 μS/cm to 2000 mS/cm | |
-30°C to +130°C | |
0.1°C | |
7 in. Color TFT | |
PC, Printer, USB Stick | |
195 x 205 x 65 mm | |
1.9 lb. | |
1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.