Learn More
Fisherbrand™ Conductivity Probe, Epoxy Body, K=1.0
Optimize the performance of your conductivity and pH/ORP meters with the Conductivity Probe, Epoxy Body, K=1.0.
Brand: Fisherbrand™ 19601-28
This item is not returnable.
View return policy
Description
- Get accurate, reliable measurements with these replacement probes and adapters.
- Features a conductivity range of 1.0 to 200,000 microsiemens
- Suitable for use with 19601-06 meter
Specifications
Conductivity Probe | |
None | |
Traceable High Accuracy, Benchtop Conductivity Meter and Probe with Calibration (19601-06) |