missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity solution 200,000μS.cm-1, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
141.00 € - 237.00 €
Specifications
Type | Conductivity Solution |
---|---|
Conductivity | 200,000μS/cm |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12914515
|
Fisher Chemical
CTE 017 500 |
500 mL | Bottle |
141.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12904515
|
Fisher Chemical
CTE 017 1L |
1000 mL | Bottle |
237.00 €
1000mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Conductivity Solution | |
200,000μS/cm |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title