missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Solution, 500μS.cm-1, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1000 mL
Specifications
Specifications
Packaging | Bottle |
Quantity | 1000 mL |
Type | Conductivity Solution |
Conductivity | 500μS/cm at 25°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction