missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solution, 1.3μS/cm, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
6 x 500 mL
Description
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- NIST traceable
- This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the product label. The low temperature coefficient of variation, reduces measurement errors and enables non- temperature controlled use and is suitable for field work
- High accuracy ± 1% at 25°C, enables the standards to be used as calibrators and/or controls, in accordance with the most exacting industrial requirements
- The product is aqueous based, non-hazardous and accurate to a specification of ± 1%. The product because it is aqueous based, has a low temperature co-efficient of variation.
Specifications
Specifications
Description | Conductivity Standard Solution |
Packaging | Bottles |
Quantity | 6 x 500 mL |
Certificate of Analysis | Yes |
For Use With (Equipment) | Conductivity Meter |
Solvent or Matrix | Aqueous |
Conductivity | 1.3μS/cm ± 1% |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction