missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard Solutions, Hanna Instruments™
A premium quality conductivity standard that is prepared against a NIST traceable potassium chloride solution.
68.45 € - 200.00 €
Specifications
Type | Calibration Standard |
---|---|
For Use With (Application) | Laboratory and Industrial Use |
Product Code | Brand | Quantity | Conductivity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Conductivity | Price | Quantity & Availability | ||||
10309960
|
Hanna Instruments
HI70030P |
20 mL | 12,880 μS/cm |
200.00 €
Pack of 25 |
In Stock
Log in to see stock. |
||||
10441141
|
Hanna Instruments
HI70031P |
20 mL | 1413 μS/cm |
68.45 €
Pack of 25 |
In Stock
Log in to see stock. |
||||
12776955
|
Hanna Instruments
HI7031 |
450 mL | 1413 μS/cm |
145.00 €
450mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12786955
|
Hanna Instruments
HI-7033 |
450 mL | 84 μS/cm | N/A | |||||
11514327
|
Hanna Instruments
HI7031+COA |
500 mL | 1413 μS/cm |
200.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11574327
|
Hanna Instruments
HI7033+COA |
500 mL | 84 μS/cm |
174.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A choice of ready mixed calibration standards traceable to NIST
- Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air.
- A premium quality 1,413 μS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution
- Temperature chart of the actual μS/cm value at various temperatures printed on each bottle
- Air tight bottle with tamper-proof seal to ensure quality
- Standardized with a conductivity meter that has been calibrated with potassium chloride NIST 2202 SRM (Standard Reference Material) in deionized water for analytical use in accordance with ISO 3696/BS3978
Specifications
Calibration Standard | |
Laboratory and Industrial Use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title