Learn More
Conductivity Standard, NIST Traceable, 1,413μS/cm, Reagecon™
Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 1,413μS/cm is an aqueous-based, non-hazardous standard with a high degree of accuracy, low temperature variation and guaranteed stability through the expiration date, even after opening the bottle.
Brand: Reagecon CSKCL
17.33 EUR valid until 2025-02-13
Use promo code "24126" to get your promotional price.
Description
Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 1,413μS/cm is an aqueous-based, non-hazardous standard with a high degree of accuracy, low temperature variation and guaranteed stability through the expiration date, even after opening the bottle.
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
- Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
- Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
- Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
- High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Specifications
500 mL | |
Conductivity Meter | |
1,413μS/cm (±1% at 25°C) |
Yes | |
Aqueous | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.