missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oakton™ Conductivity and TDS Standard
Oakton™ conductivity and TDS standards are traceable to NIST standard reference materials and ideal for laboratory and field analysis.
Brand: Oakton™ 00653-32
14.42 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- ISO 9000 and NIST Treaceable
- Helps calibrate conductivity instruments to ensure accuracy
- Standard values give at 25°C
- Use when readings are expected to be around 80 mS (80,000 μS/cm)
- KCI, NaCI and 442 conversions, and temperature corrections printed on bottle for ease of use (excluded on 60 mL bottle, but included in insert)
- Recommended four month shelf life once opened
Specifications
Conductivity and TDS Standard | |
80,000 μS/80 mS | |
Calibration of Conductivity Instrument |
500 mL | |
ISO 9000 quality guideline; traceable to NIST standards |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.