Learn More
Quickfit™ Single Cone with Long Tip
Allows cone to be joined to glass tubing
Brand:  Quickfit CBB34UB
 View more versions of this product
Product Code. 15557760
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Useful for construction of stirrer guides
  • Unbadged and free from ceramic markings
  • Avoids problem of badge charring at high temperatures which can give an untidy appearance
  • Long tip cones manufactured with minimum shank length of 90mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10Pack
Single Cone with Long Tip
34/35
2mm