Learn More
Quickfit™ Single Cone with Long Tip
Allows cone to be joined to glass tubing
Brand: Quickfit CBB45UB
This item is not returnable.
View return policy
Description
- Useful for construction of stirrer guides
- Unbadged and free from ceramic markings
- Avoids problem of badge charring at high temperatures which can give an untidy appearance
- Long tip cones manufactured with minimum shank length of 90mm
Specifications
10/Pk. | |
Single Cone with Long Tip | |
2.3mm |
Dia. Shank: 42mm, Bore: 31mm | |
45/40 |