Learn More
Lenz Laborglasintrumente Cones with Tip
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100334
 View more versions of this product
Product Code. 10532681
  • 134.00 € / Pack of 10
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12249 form G and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
2 mm
Borosilicate Glass (3.3)
NS 34/35
30 mm