missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Conical Dropping Funnels with PTFE Plug
Constructed from borosilicate glass. Lenz Laborglasinstrumente™ Conical Dropping Funnels with PTFE Plug are utilized in liquid-liquid extractions to separate a mixture into immiscible solvent phases of different densities.
88.00 € - 99.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11876094
|
Lenz Laborglasinstrumente™
04474904 |
250 mL |
99.00 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
11886114
|
Lenz Laborglasinstrumente™
04472802 |
50 mL |
88.00 €
Each |
Estimated Shipment: 27-12-2024
Log in to see stock. |
|||||
Description
- Conical shaped with NS socket and NS cone
- With PTFE plug
- With screwthread retaining device
- With inscription label, without scale
Specifications
Borosilicate Glass | |
Dropping Funnel |