Learn More
Fisherbrand™ Connecting Cables
Connecting cable with BNC Plug for combined ion-sensitive electrode with solid membranes that is ideal for measuring agriculture, water analysis, and calibration of ionic electrodes.
Brand:  Fisherbrand™ 9001041-003B
Product Code. 11746348
  • 59.45 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Connecting Cable with BNC Plug
  • For combined ion-sensitive electrode with solid membranes
  • For use with measuring agriculture, water analysis, and calibration of ionic electrodes
Specifications
Specifications
1000 ppm
Connecting Cable
Electrolyte Solution Ring
BNC
Agriculture and Water Analysis, Calibration of Ionic Electrodes
0°C to 50°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Connecting Cables > Connector Type: BNC; Includes: Electrolyte Solution Ring