missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Constans Safety Shoes

Developed for semiorthopedic adjustments requiring customized in-soles.

401.00 €

Specifications

Color Black
For Use With (Application) semiorthopedic adjustments requiring customized in-soles
Liner Material Nappa leather
Material Leather
Sole Features Woven zero-penetration insole, Heat-resistant outsole
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
10173974
View Documents
Emma Safety Footwear
MM13207046
46
401.00 €
1 pair
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
15287038
View Documents
Emma Safety Footwear
MM13207040
40
401.00 €
1 pair
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
Description

Description

  • Full grain leather upper construction
  • Nappa leather lining
  • With sole plate for wet work
  • Woven zero penetration in-sole
  • Antibacterial inlay sole
  • Vibram™ Nitril / HRO outsole
Specifications

Specifications

Black
Nappa leather
Woven zero-penetration insole, Heat-resistant outsole
Constans Safety Shoes
semiorthopedic adjustments requiring customized in-soles
Leather
Vibram™ Nitrile
Videos
SDS
Documents

Documents

Product Certifications