missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Door for Climate Chamber, Stainless Steel, with Safety Lock

Door with lock and key (Safety Lock).

Brand:  Memmert™ HPP750B6

Product Code. 15847191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Door with one lock.
This item is not returnable. View return policy

Description

Description

This door is for use with the Memmert Constant Climate Chamber.

Memmert Constant Climate Chamber:
  • Intuitive and easy-to-use operating menu
  • temperature and humidity ramps programmable via AtmoCONTROL software
  • almost exclusive use of high-quality, corrosion-resistant and easily cleanable stainless steel for the working chamber and housing
  • Peltier heating and cooling concept specifically adapted for a precise and homogenous temperature control
  • a wide range of options for programming and documentation using interfaces, integrated data logger and software AtmoCONTROL
Specifications

Specifications

749 L
Textured Stainless Steel
15°C to 40°C
HPP750B6
Stainless Steel
10 to 90%
Door with Lock
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.