Learn More
Liofilchem™ Contact Slide 2
Rose Bengal CAF Agar and Neutralizing / PCA and TTC and Neutralizing Flex Dip-slide with a selective medium for detection of yeasts and moulds and a non selective medium for total bacterial count.
47.81 €
Specifications
Manufacturing Origin | Italy |
---|---|
Sterility | Non-sterile |
Target Organism Class | Eukaryotic Microorganism, Fungi, Gram-positive Organisms, Fungi |
Description
- Contact Slide 2 is a ready-to-use device with two different media coated onto a plastic support used for the microbial monitoring of surfaces and liquids even in the presence of residues of disinfectants. The selective medium allows the isolation and enumeration of yeasts and moulds . The other medium is used for enumeration of bacteria.
Specifications
Italy | |
Non-sterile | |
Eukaryotic Microorganism, Fungi, Gram-positive Organisms, Fungi |
Safety and Handling
missing translation for 'shelfLife' : 9 Months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.