missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Container with Integrated Lid

Container with Integrated Lid integrates crocodile lid, ideal for catering. Stackable when closed and nesting when the lid is open.

58.80 € - 109.00 €

Specifications

Type Storage Tote
Material HDPE
Color Blue
Shape Rectangular
View More Specs

Products 2
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Price Quantity & Availability  
17948661
View Documents
Gilac™
G181338
25 L 400 x 300 x 310 mm
58.80 €
Each
Please to purchase this item. Need a web account? Register with us today!
17928731
View Documents
Gilac™
G181438
55 L 600 x 400 x 315 mm
109.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Container with integrated ''crocodile'' lid.
  • Ideal for catering.
  • Stackable when closed and nesting when the lid is open.
Specifications

Specifications

Storage Tote
HDPE
Blue
Rectangular
Videos
SDS
Documents

Documents

Product Certifications