missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Containers with Sloping Base
This range of containers with sloping bases ensures that material can be completely drained from within and includes a ferrule at the base for easy emptying. These containers have a mirror polish finish for easy cleaning.
Brand: Sampling Systems™ A616-200
This item is not returnable.
View return policy
Description
- Made from 316L stainless steel
- Includes lid, clamp and silicone seal
- Other seals available
- Handles enable easy transportation
Specifications
Drum | |
200 L | |
Liquid Storage |
52.8 gal. | |
Silver | |
Stainless Steel (316L) |