missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Amplitude™ Epsilon™ Cleanroom Wipes
BIOPHARMAGREENER_CHOICE

Equip workers with the tools to keep cleanrooms clean. The Amplitude™ Epsilon™ Cleanroom Wipes, are a durable solution for ISO Class 6 applications.

385.00 €

Specifications

Product Type Cleanroom Wipe
Material Polyester Cellulose
Certifications/Compliance ISO Class 6
Color Blue
For Use With (Application) Cleanroom
View More Specs

Products 1
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
16635514
BIOPHARMAGREENER_CHOICE
View Documents
Contec™
AMEP0001
9 x 9 in. (22.86 x 22.86 cm)
385.00 €
Pack of 3600
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
 
Description

Description

  • Polyester/cellulose hydroentangled nonwoven wipes are strong and durable and provide improved cleanliness
  • Heavyweight, with high sorbency
  • Dye-fast blue color for differentiation between areas of use
  • Excellent all-purpose cleanroom wipes
  • Lot-numbered for traceability
  • Made of hydroentangled premium polyester/cellulose for improved cleanliness
  • Excellent for wiping and absorbing spills and in general purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical device, and printed circuit board industries
  • Double packed to preserve product integrity

Recommendation: Test colorfastness with each process before use.

Order Info

Basis weight, 75g/m2.

Specifications

Specifications

Cleanroom Wipe
ISO Class 6
Cleanroom
Polyester Cellulose
Blue
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Contec™ Amplitude™ Epsilon™ Cleanroom Wipes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.