missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ CONSTIX™ Surface Validation Swabs
Contec™ CONSTIX™ Surface Validation Swabs designed for the evaluation of surface cleaning protocols in biopharmaceutical industries.
Brand: Contec™ SV-2
9.29 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Package contains five polyester swabs specially processed to minimize background TOC contribution from the swab
- Low background TOC content for minimal interference with test results
- Large surface of the swab head and the long flexible handle provide exceptional surface coverage when swabbing
- Strong, durable polyester swabs for optimum retrieval
- Sterilizable by most common methods
- Autoclavable
- Swabs sealed in nylon pouch
Specifications
0.12 in. | |
Evaluation of surface cleaning protocols in bio-pharmaceutical industries | |
3 mm | |
5.2 in. | |
0.8 in. | |
6.3 mm | |
6.4 mm | |
Ultrasonically Sealed | |
Knit Polyester |
6 in. | |
Swab | |
152.4 mm | |
13.2 mm | |
2 cm | |
0.23 in. | |
0.25 in. | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction