missing translation for 'onlineSavingsMsg'
Learn More

Contec™ EasyReach™ Cleaning Pads
BIOPHARMAGREENER_CHOICE

EasyReach™ Cleaning Pads are very absorbent, which ensures even coverage of disinfectant or IPA.

Brand:  Contec™ MEQT0002SZ

266.99 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 17143634

  • 368.00 € / Pack of 60
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Single-use sealed polyester
  • Provide a low level of particulates which is essential for use in product contact areas
  • Very absorbent, ensuring good even coverage of disinfectant or IPA
  • Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame
  • Sterile cleaning pads are presaturated with 70% IPA
Specifications

Specifications

Cleaning Pad
19 cm
7 cm
Not Autoclavable
Single-use
Cleaning Isolators, LAF Hoods, Glove Boxes and Other Mini Environments
Sterile
7.5 in.
2.8 in.
Quilted Polyester
White
7.5 x 2.8 in. (19 x 7 cm)
4/Bag, 4 Pads/Bag, 15 Bags/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers