missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polynit™ Heatseal Wipes

Clean the exterior of products and cleanroom surfaces. The Polynit™ Heatseal Wipes - ISO Class 4 are a 100% textured polyester with laser-cut sealed edges to prevent dust and residue. Extremely low in particles, fibers and non-volatile residues (NVRS).

Brand:  Contec™ PNHS-44

731.50 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


Product Code. 15323346

  • 1050.00 € / Pack of 300
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Textured 100% polyester no-run knit wipes with heatsealed edges
  • Laser-cutting seals fibers at wipe's edge and allows for 100% utilization of wipe
  • Laundered and packaged in a ISO Class 4 cleanroom
  • Extremely low in particles, fibers and non-volatile residues (NVRS)
  • Good sorbency with solvents
  • Abrasion and chemical resistant
Specifications

Specifications

Polynit Heatseal Wipes
10.16 cm
10.16 cm
140 g/m2
4 x 4 in. (10.16 x 10.16 cm)
300 Wipes/Bag
4 in.
4 in.
Polyester
ISO Class 4
Ideal for general purpose cleaning
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.