missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polynit™ Wipes
BIOPHARMAGREENER_CHOICE

Clean surfaces of products and process tools. The Polynit™ Wipes are a durable option for ISO Class 5 environments.

Brand:  Contec™ PN-99

422.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16631284

This item is not returnable. View return policy

Description

Description

  • For general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment

Order Info

Laundered and packaged in an ISO Class 4 cleanroom. Available sterile per AAMI ISO Method I Guidelines with a SAL of 10-6, packaged in peelable outer bags with linear tear inner bags.

Specifications

Specifications

Wipe
23 cm
23 cm
Not Autoclavable
White
23 x 23 cm (9 x 9 in.)
150 Wipes/Bag (Flat Stacked)
Non-sterile
9 in.
9 in.
Polyester
ISO Class 5
Single-use
Cleanroom
2-ply
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers