Learn More
Contec™ ProZorb™ Nonwoven Cleanroom Wipes
Manufactured with cellulose and polyester through a special process that creates apertures in the fabric while enhancing the thickness. Contec™ ProZorb™ Nonwoven Cleanroom Wipes provide a textured surface with excellent sorptive capacity and particle removal capability.
474.00 €
Specifications
Product Type | Cleanroom Wiper |
---|---|
Material | Polyester Cellulose |
Basis Weight | 55 |
Certifications/Compliance | ISO Class 5 |
For Use With (Application) | Cleanroom |
Description
- Textured polyester/cellulose nonwoven wipes promotes effective cleaning
- Low levels of particles and extractables
- Strong and durable
- Designed for nonabrasive applications in the biotech, pharmaceutical, electronics, semiconductor industries and more
- Recommended for use in ISO Class 5 cleanroom applications; Use for product wipe down prior to cleanroom entry
- Great for spill clean up in critical environments
- No chemical binders or surfactants
Order Info
Basis weight: 55g/m2.
Specifications
Cleanroom Wiper | |
55 | |
Cleanroom |
Polyester Cellulose | |
ISO Class 5 |