Learn More
Contec™ QuickConnect™ Mop Handles and Hooks
Choose from a lightweight anodized aluminum, durable electropolished stainless steel or fiberglass. QuickConnect™ Mop Handles are available in fixed length or telescoping models to fit all mopping needs.
24.65 € - 471.00 €
Product Code | Brand | Product Type | Material | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Material | Length (English) | Price | Quantity & Availability | |||
11952405
|
Contec™
2644 |
2 Piece Extendable Handle | Stainless Steel | 16 to 30 in. |
263.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
11932405
|
Contec™
2642 |
2 Piece Extendable Handle | Anodized Aluminum | 16 to 30 in. |
122.00 €
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||
11942405
|
Contec™
2643 |
2 Piece Extendable Handle | Anodized Aluminum | 50 to 92 in. |
157.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
11962405
|
Contec™
2645 |
2 Piece Extendable Handle | Stainless Steel | 50 to 92 in. |
326.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
11932415
|
Contec™
2686 |
1 Piece Fixed Length Handle | Stainless Steel | 16 in. |
24.65 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
15805597
|
Contec™
2746 |
2 Piece Extendable Handle | Fiberglass | 71 to 130 in. |
116.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
11972415
|
Contec™
2725E |
1 Piece Fixed Length Handle | Stainless Steel | 60 in. |
70.80 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
|||
16031012
|
Contec™
2746S |
2 Piece Extendable Handle | Fiberglass | 37.5 to 65.5 in. |
122.00 €
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17958312
|
Contec™
CFH334 |
Extendable Handle | Carbon Fiber | 69 to 334 in. |
471.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17918312
|
Contec™
CFH114 |
Extendable Handle | Carbon Fiber | 63 to 114 in. |
178.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17908312
|
Contec™
CFH32 |
Extendable Handle | Carbon Fiber | 18 to 32 in. |
118.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17948312
|
Contec™
CFH70 |
Extendable Handle | Carbon Fiber | 40 to 70 in. |
122.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17938312
|
Contec™
CFH224 |
Extendable Handle | Carbon Fiber | 66 to 224 in. |
336.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Choose from a lightweight anodized aluminum, durable electropolished stainless steel or fiberglass. Contec™ QuickConnect™ Mop Handles are available in fixed length or telescoping models to fit all mopping needs.
QuickConnect™ mop head frames and handles offer maximum flexibility for cleaning in critical environments. Invest in one handle and choose various interchangeable heads and frames for specific applications.
QuickConnect Mop Handles:- Use with full Contec™ line of mopping systems
- Autoclavable
- Telescoping lengths: 16 in. (41 cm) extends to 30 in. (76 cm), 50 in. (127 cm) handle extends to 92 in. (234 cm), 69 in. (175 cm) extends to 10.8 ft. (3.3 m)
- Fixed lengths: 16 in. (41 cm), 60 in. (152 cm)
- For hard to reach ceilings and walls, a 16 ft. (4.9 m) anodized aluminum handle is available; 4 piece handle easily separates for autoclaving
- QuickConnect handle hook, available to fit 0.9 in. (2 cm) and 1 in. (2.5 cm) handles
- Mop handle hook securely holds QuickConnect handles to the bucket trolleys, reducing the chance for mishaps or spills
Specifications
Non-sterile | |
Autoclavable |