missing translation for 'onlineSavingsMsg'
Learn More

Contec™ QuickTask™ Flat Hinged Mop Head Frames
BIOPHARMAGREENER_CHOICE

Contec™ QuickTask™ Flat Hinged Mop Head Frames are durable, lightweight and exclusively designed for the Contec™ QuickTask™ Mop System.

Brand:  Contec™ 2740

35.24 EUR valid until 2025-03-09
Use promo code "24130" to get your promotional price.


 View more versions of this product

Product Code. 15895587

  • 53.05 € / Each
Estimated Shipment: 30-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in stainless steel or blue polypropylene
  • Autoclavable
  • Compatible with most common disinfectant solutions and solvents
  • The QuickTask™ system consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly
  • The QuickTask™ system is ideally suited for cleaning floors, including perforated cleanroom floors and grates
TRUSTED_SUSTAINABILITY
Specifications

Specifications

41 x 13 cm (16 x 5 in.)
White
Non-sterile
Reusable
Ideally suited for cleaning and applying disinfectant to ceilings, walls and floors including perforated cleanroom floors and grates
Polypropylene
Mop Head Frame
Blue
Autoclavable
Contec™ QuickConnect™ 60 in. Stainless Steel Handle (Fisher cat. no. 19168798)
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.