missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Continuous Bottle-Top Burette

Ensure precise and continuous titration. VITLAB™ Continuous Bottle-Top Burette features a digital control panel with hand wheels to easily control titration rate and calibration. Adjustable components and a double-piston pump ensure precise results and safe operation. Available in two volume rates.

Specifications

Accuracy 0.2 %
Graduations 0.01 (up to 99.99 mL) mL, 0.1 (100 to 999.9 mL) mL
For Use With (Application) Continuous Titration of a Range of Media for Chemical Volumetric Analysis
Includes (2) 1.5 V Microbatteries, GL 45 Connecting Threads and GL 32, GL 38 and S*40 Size PP Thread Adapters, Instruction Manual and Quality Certificate, Telescopic Discharge Tube (140-220 mm), Telescopic Filling Tube (200-350 mm)
Precision 0.1 %
View More Specs

Description

Description

Chemical volumetric analysis with minute liquid volumes requires substantial concentration. The VITLAB™ Continuous Bottle-Top Burette enables continuous titration up to a concentration of 1 mol/L, which leads to rapid, convenient and accurate results.

Digital display panel simplifies operation
  • Angled display shows titration volume in large, easily read numbers
  • Easy-to-use, medium-specific calibration using the keypad buttons
  • Precise control of the titration rate by rotating two large, easy-grip hand wheels
  • Also includes battery, pause and mode indicators and function keys/buttons
Adjustable, continuous titration control
  • Patented double-piston pump supplies the titration in a continuous, pulse-free manner
  • Telescopic discharge tube can be adjusted (height and length) for use with short or tall bottles
  • Rotates freely 360° around the bottle for optimal orientation of the label
  • Adjustable for various bottle sizes using the corresponding thread adapter
  • Long operating life using two easily-replaced 1.5 V microbatteries
Quality and safety assured
  • Innovative recirculation valve prevents the loss of reagents and reduces splashes
  • Compact design with low center of gravity reduces risk of overturning, especially with smaller bottles
  • Fulfils all requirements for test equipment monitoring without instrument downtime
  • Margins of error are under the DIN EN ISO 8655-3 standard, even for partial volumes
Specifications

Specifications

0.2 %
Continuous Titration of a Range of Media for Chemical Volumetric Analysis
0.1 %
DIN 12600, DIN EN ISO 8655-3
0.01 (up to 99.99 mL) mL, 0.1 (100 to 999.9 mL) mL
(2) 1.5 V Microbatteries, GL 45 Connecting Threads and GL 32, GL 38 and S*40 Size PP Thread Adapters, Instruction Manual and Quality Certificate, Telescopic Discharge Tube (140-220 mm), Telescopic Filling Tube (200-350 mm)
Continuous Buret
Videos
SDS
Documents

Documents

Product Certifications