missing translation for 'onlineSavingsMsg'
Learn More

SymLine Continuous Level Control Accessories

Accessories for Continuous Level Control

Brand:  SymLine 108009

41.50 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10809340

  • 52.55 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications

Specifications

Grounding cable, 2 x clamp, L = 1,5 m
2 x clamps
Grounding Cable
1.5 m
Safety Waste Caps and Connection Rings Funnels
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers