Learn More
SymLine Continuous Level Control Accessories
Accessories for Continuous Level Control
Brand:  SymLine 108291
333.31 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17317429
  • 595.00 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications
Specifications
Capacitive disc sensor, housing material: stainless steel, Active surface: PTFE, operating distance 1 - 10 mm adjustable, output function: NAMUR. Switch amplifier necessary (108278)
ATEX zone 0, ATEX Ex II 1 G Ex ia IIC T1-T6 Ga
Capacitive Disc Sensor